Masterflex Leak Detector Sensor w/ Plastic Catch Basin
Representative image only.
Masterflex Item # WW-77097-12
Sensors shut down the pump drive in less than 1 second in the event of fluid leak or tubing failure
  • Options available for mounting and connecting sensors to match your equipment and application
  • Sensor 77097-12 includes a plastic catch basin; use where a larger sensing area is needed and for fluids with low conductivity
PRODUCT OPTIONS
₹53,900.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionLeak Detector Sensor w/ Plastic Catch Basin
Key Features
  • Options available for mounting and connecting sensors to match your equipment and application
  • Sensor 77097-12 includes a plastic catch basin; use where a larger sensing area is needed and for fluids with low conductivity