Masterflex Fitting, PVDF Body with Polypropylene Insert, Female Luer Lock x Three-Way Stopcock
Representative image only.
Masterflex Item # WW-40619-99
Build leakproof fluid handling systems
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
₹8,500.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Outlet PortFemale Luer Lock
  • Inlet PortFemale Luer Lock
  • Wetted MaterialsNatural PVDF, Polypropylene
  • Flow PatternThree-Way
  • DescriptionFitting, PVDF Body with Polypropylene Insert, Female Luer Lock x Three-Way Stopcock
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Designed for quick and easy assembly with flexible tubing
More About this Item
These one-way stopcocks feature an all-black PVDF construction.