Masterflex 316 SS Metering Valve, 90 deg flow, 1200 mL/min
Representative image only.
Masterflex Item # WW-03218-83
Choose from different metal materials based on your application
PRODUCT OPTIONS
₹18,200.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))0.125
  • Outlet Port1/8" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port1/8" NPT(F)
  • Orifice (in)0.082
  • Orifice (mm)2.0828
  • Max Flow Rate For Air (mL/min)220000
  • Max Flow Rate For Water (mL/min)1200
  • Max Temperature (° F)250
  • Max Temperature (° C)121
  • Wetted MaterialsStainless Steel
  • Flow Pattern90°
  • Description316 SS Metering Valve, 90 deg flow, 1200 mL/min
More About this Item
Brass valves feature Buna N O-rings; stainless steel valves have Viton® O-rings.