Marvel Scientific MS24RFSHRW 24" Hazardous Location Refrigerator/Freezer, 6.1 cu ft, 120V

Representative image only.
Marvel Scientific Mfr # MS24RFSHRW – Item # WW-44766-04
Refrigerator/Freezers that are appropriate for noncritical temperature storage applications
  • Listed by Underwriters Laboratories for Class 1, Division 1 and 2, Group C and D applications
  • Wiring complies with National Electrical Code, Article 501-4 for Class 1, Division 1 and 2
  • Complies with ANSI/UL 1203 Standard, NFPA 70 and NFPA 99
  • 1 year warranty parts & labor

SPECS & DESCRIPTION

Specifications & Description

  • Refrigerator Capacity (cu ft)6.1
  • Refrigerator Capacity (Liters)173
  • Freezer Capacity (cu ft)0.7
  • Freezer Capacity (Liters)20
  • Min Temperature - Refrigerator (° F)33
  • Max Temperature - Refrigerator (° F)45
  • Min Temperature - Refrigerator (° C)1
  • Max Temperature - Refrigerator (° C)7
  • Min Temperature - Freezer (° F)0
  • Max Temperature - Freezer (° F)15
  • Min Temperature - Freezer (° C)-18
  • Max Temperature - Freezer (° C)-9
  • Temperature ControlMechanical Thermostat
  • Refrigerator Interior Width (in)21
  • Refrigerator Interior Height (in)28
  • Refrigerator Interior Depth (in)20 3/4
  • Refrigerator Interior Width (cm)53
  • Refrigerator Interior Height (cm)71
  • Refrigerator Interior Depth (cm)53
  • Exterior Width (in)23 7/8
  • Exterior Height (in)34
  • Exterior Depth (in)24
  • Exterior Width (cm)61
  • Exterior Height (cm)86
  • Exterior Depth (cm)61
  • Number of Doors1 solid
  • Number of Shelves2 interior, 3 door
  • Power (VAC)115
  • Power (Hz)60
  • Power (amps)3.3
  • Description24\" Hazardous Location Refrigerator/Freezer, 6.1 cu ft, 120V
  • Warranty1 year parts & labor
1 Year Parts and LaborCanadian Standards Association

Key Features

  • Listed by Underwriters Laboratories for Class 1, Division 1 and 2, Group C and D applications
  • Wiring complies with National Electrical Code, Article 501-4 for Class 1, Division 1 and 2
  • Complies with ANSI/UL 1203 Standard, NFPA 70 and NFPA 99
  • 1 year warranty parts & labor

More About this Item

Store heat-sensitive materials in these space-efficient refrigerators. All models have quiet, oversized refrigeration units to enable quick recovery from door openings. CFC-free refrigerant. Recessed thermostat enables flexible control. Doors are easily changed to left-side hinging.

This Hazardous Location (Explosion Proof) combo refrigerator/freezer's construction features include a specially designed compressor, overload, relay and thermostat system with encclosed electrical components that prevent accidental ignition of flammable vapors that may be present. A small amount of dedicated freezer space is created by utlizing a “U” shaped plate evaporator across the top of the interior and isolating it from the main body with a spring-closing interior door and thermal divider. The remainder of the interior is refrigerator storage space. Other features include, manual defrost, three interior door shelves, top edge mounted door handle, and 4 leveling legs for easy height adjustment. Additional ventilation clearance is needed if the unit is being installed under lab benches or within casework.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat