Marvel Scientific 6CRF-GLD General Purpose Undercounter Refrigerator/Freezer with Glass Door, 6 cu ft
Representative image only.
Marvel Scientific Mfr # 6CRF7104 – Item # WW-44765-46
Refrigerator/Freezers that are appropriate for noncritical temperature storage applications
  • 1 year warranty parts & labor
SPECS & DESCRIPTION
Specifications & Description
  • Refrigerator Capacity (cu ft)5.8
  • Refrigerator Capacity (Liters)165
  • Min Temperature - Refrigerator (° F)33
  • Max Temperature - Refrigerator (° F)42
  • Min Temperature - Refrigerator (° C)1
  • Max Temperature - Refrigerator (° C)6
  • Number of Doors1 glass
  • Number of Shelves2 interior, 3 door
  • Power (VAC)115
  • Power (Hz)60
  • DescriptionGeneral Purpose Undercounter Refrigerator/Freezer with Glass Door, 6 cu ft
  • Warranty1 year parts & labor
1 Year Parts and LaborCanadian Standards AssociationUL Listed.
Key Features
  • 1 year warranty parts & labor
More About this Item
Store heat-sensitive materials in these space-efficient refrigerators. All models have quiet, oversized refrigeration units to enable quick recovery from door openings. CFC-free refrigerant. Recessed thermostat enables flexible control. Doors are easily changed to left-side hinging.
General Purpose Models are ideal for basic storage of chemicals and reagents. These units are available in undercounter and large capacity models.