Mark-10 MR52-20Z Torque Sensor, 20 ozFin / 1400 gFcm / 14 kgFmm / 140 Nmm / 14 Ncm
Representative image only.
Mark-10 Mfr # MR52-20Z – Item # WW-25302-10
Rugged aluminum housing
  • Universal mounting
  • Used for torque tool calibration or as a component of a torque testing system
  • Rugged aluminum housing
  • Universal mounting
  • Used for torque tool calibration or as a component of a torque testing system
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (oz-in)20
  • Capacity (g-cm)1400
  • Resolution (oz-in)0.01
  • Resolution (g-cm)1
  • Resolution (N-cm)0.01
  • Accuracy±0.35% of full scale + indicator
  • DescriptionPlug & Test Bidirectional Static Torque Sensor; 14 N
  • Warranty3 year
3 Year WarrantyConformite Europeenne
Key Features
  • Universal mounting
  • Used for torque tool calibration or as a component of a torque testing system
  • Rugged aluminum housing
  • Universal mounting
  • Used for torque tool calibration or as a component of a torque testing system
  • 3 year warranty
More About this Item
Calibrate torque tools with this torque measurement sensor, or use it as a component of a torque testing system. Tapped holes on every surface allow for easy mounting to a work bench, or fixture within a testing system.
Table Mounting Kit and 3/8" Jacob's chuck available separately.
Calibrate torque tools with this torque measurement sensor, or use it as a component of a torque testing system. Tapped holes on every surface allow for easy mounting to a work bench, or fixture within a testing system.
Table Mounting Kit and 3/8" Jacob's chuck available separately.