Mark-10 MR52-12 Torque Sensor, 12 lbFin / 1 lbFft / 192 ozFin / 135 Ncm / 1.35 Nm
Representative image only.
Mark-10 Mfr # MR52-12 – Item # WW-25302-12
Rugged aluminum housing
  • Universal mounting
  • Used for torque tool calibration or as a component of a torque testing system
  • Rugged aluminum housing
  • Universal mounting
  • Used for torque tool calibration or as a component of a torque testing system
  • 3 year warranty
PRODUCT OPTIONS
₹198,000.00INR / Each
₹40,000.00 - ₹46,800.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (oz-in)192
  • Capacity (lb-in)12
  • Resolution (oz-in)0.1
  • Resolution (lb-in)0.005
  • Resolution (N-cm)0.1
  • Accuracy±0.35% of full scale + indicator
  • DescriptionPlug & Test Bidirectional Static Torque Sensor; 135 N
  • Warranty3 year
3 Year WarrantyConformite Europeenne
Key Features
  • Universal mounting
  • Used for torque tool calibration or as a component of a torque testing system
  • Rugged aluminum housing
  • Universal mounting
  • Used for torque tool calibration or as a component of a torque testing system
  • 3 year warranty
More About this Item
Calibrate torque tools with this torque measurement sensor, or use it as a component of a torque testing system. Tapped holes on every surface allow for easy mounting to a work bench, or fixture within a testing system.
Table Mounting Kit and 3/8" Jacob's chuck available separately.
Calibrate torque tools with this torque measurement sensor, or use it as a component of a torque testing system. Tapped holes on every surface allow for easy mounting to a work bench, or fixture within a testing system.
Table Mounting Kit and 3/8" Jacob's chuck available separately.