Marathon Stopwatch, 1-button, NIST Calibration Report
Representative image only.
Marathon Mfr # ST211003-W/NIST – Item # WW-08758-04
Precision movements are antimagnetic and shock resistant
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Timing Capacity30 minutes
  • Resolution0.2 sec, 0.01 minute divisions
  • Sweep (seconds)60
  • MovementLever
  • CaseNickel-chrome plated
  • CertificationsNIST-traceable
  • LensShatterproof Plexiglass
  • Depth (in)3 1/2
  • Width (in)1 3/4
  • Depth (cm)8.9
  • Width (cm)4.4
  • DescriptionStopwatch, 1-button, NIST Calibration Report
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Stopwatches feature shock absorbers on the balance wheel, temperature-compensated hairspring, and unbreakable mainsprings. Easy-to-read numbers are clearly visible on the large, 1-3/4"-diameter face. All watches have locked returns to prevent accidental reset to zero.
One-Button stopwatches feature fifteen jewel movement and a single button that starts, stops, and resets the watch. Shatterproof Plexiglas® lens offers greater scratch and chemical resistance. Includes an NIST-traceable calibration report supplied by the manufacturer.