Marathon Septa, CenterGuide™, 9 mm; 50/Pk
Representative image only.
Marathon Mfr # 239779 – Item # WW-01999-00
Long injection life, pre-pierced for minimal coring
  • CenterGuide™ design requires less force for initial penetration
  • 1 month warranty
PRODUCT OPTIONS
₹18,400.00INR / Pkg of 50
SPECS & DESCRIPTION
Specifications & Description
  • Diameter (mm)9
  • DescriptionSepta, CenterGuide, 9 mm; 50/Pack
  • Warranty1 month
1 Month Warranty
Key Features
  • CenterGuide™ design requires less force for initial penetration
  • 1 month warranty
More About this Item
Marathon™ Septa have outstanding durability in either autosampler or manual injection applications. Operate at a maximum temperature of 400°C and achieve up to 400 injections per septum. Pre-piercing minimizes injection coring on select CenterGuide septa.