Marathon™ Septa have outstanding durability in either autosampler or manual injection applications. Operate at a maximum temperature of 400°C and achieve up to 400 injections per septum. Pre-piercing minimizes injection coring on select CenterGuide septa.