MadgeTech OctRTDTempV2 8-Channel RTD Temperature Recorder; Screw Terminals
Representative image only.
MadgeTech Mfr # OctRTDTempV2 – Item # WW-18003-08
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Number of Channels8
  • Input TypeTemperature (eight external)
  • Max Temperature (° F)1562
  • Min Temperature (° F)-328
  • Min Temperature (° C)-200
  • Max Temperature (° C)850
  • Description8-Channel RTD Temperature Recorder; Screw Terminals
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
This is an all-in-one compact, portable, easy to use device that will measure and record up to thousands of measurements per channel. The storage medium is non-volatile, solid-state memory - providing maximum data security even if the battery becomes discharged. The device can be started and stopped directly from your computer and its small size allows it to fit almost anywhere. Simply plug it into an empty COM port and our user-friendly software does the rest.
FEATURES
  • Type J, K, T, E, R, S, B, N Thermocouple or 100 Ohm RTD models
  • Real-time operation
  • Low cost
  • User-friendly
  • Battery powered - user replaceable
  • Programmable start time
  • Small size
  • Non-volatile solid state memory
  • Readings selectable 1 every 5 sec to 1 every 12 hours