Lovibond 433280 Comparator 2000, Hazen/AlPHa Low
Representative image only.
Lovibond Mfr # 433280 – Item # WW-99562-44
Economical, visual color measurement in laboratory, plant, or field
  • Two-Field Comparator system for simple color matching
  • Supplied with lighting unit for standardized measurements
  • Color scales include Pt-Co, Hazen, and APHA
  • 1 year warranty
PRODUCT OPTIONS
₹1,184,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestWater and wastewater, edible oils, petroleum
  • Measurement Points0 to 70 mg Pt/L
  • DescriptionComparator 2000, Hazen/AlPHa Low
  • Warranty1 year
1 Year Warranty
Includes
Discs containing colored glass filters, sample cells/tubes, calibration filter.
Key Features
  • Two-Field Comparator system for simple color matching
  • Supplied with lighting unit for standardized measurements
  • Color scales include Pt-Co, Hazen, and APHA
  • 1 year warranty
More About this Item
These durable, portable systems allow you to make quick visual comparisons between your sample and a range of stable colored glass standards on a disc. Battery operated illuminator offers flexibility for laboratory, plant, or field color grading. With the two-section field of view, the sample transmittance is directly compared with individual colored glass standards on a test disc.
The comparator is an easy-to-use, entry level color measurement system that compares the color of the sample directly against the color of glass in a disc. Just rotate the disc to get the closest result to the scale. Ideal for light-colored samples with visual color (pathlength up to 40 mm); an illuminator is added to systems for measuring unsaturated samples requiring a longer pathlength.