Life Technologies SH30714.05 Media Bag, Sterile, 2 L, 3 Ports

Representative image only.
Life Technologies Mfr # SH30714.05 – Item # WW-06342-27
The only single-use containers that is free of any animal-derived components

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (mL)2000
  • Capacity (L)2
  • Number of Ports3
  • DescriptionMedia Bag, Sterile, 2 L, 3 Ports

More About this Item

These containers are single-use liquid handling systems engineered for biotech and pharmaceutical applications. Replace traditional rigid walled plastic and glass tubes, flasks, bottles and carboys with disposiable sterile Labtainers, minimize the risk of cross contamination, reduce cleaning and turnaround time. All products pass USP Class VI biocompatible testing. Containers are end-ported and hangable, allows for easy dispensing. No dip tubes required to evacuate solutions. No air entrapment or foaming problems. Containers maintains integrity at temperatures ranging from –80oC to 60oC. Suitable for packaging, ambient and cryogenic storage of a variety of liquids including media, buffers, process liquids, and biological products. Gamma irradiated, double polybagged.

Line 1: 3/8'' (9.5 mm) Quick Connect Insert, 12” (30.5 cm) tube length
Line 2: 3/8'' (9.5 mm) Quick Connect Body, 12” (30.5 cm) tube length
Line 3: 1/4'' (6.4 mm) Plugged, 2” (5.1 cm) tube length

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat