Life Technologies SH30712.05 Media Bag, Sterile, 2 L, 3 Ports
Representative image only.
Life Technologies Mfr # SH30712.05 – Item # WW-06343-23
The only single-use containers that is free of any animal-derived components
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)2000
  • Capacity (L)2
  • Number of Ports3
  • DescriptionMedia Bag, Sterile, 2 L, 3 Ports
More About this Item
These containers are single-use liquid handling systems engineered for biotech and pharmaceutical applications. Replace traditional rigid walled plastic and glass tubes, flasks, bottles and carboys with disposiable sterile Labtainers, minimize the risk of cross contamination, reduce cleaning and turnaround time. All products pass USP Class VI biocompatible testing. Containers are end-ported and hangable, allows for easy dispensing. No dip tubes required to evacuate solutions. No air entrapment or foaming problems. Containers maintains integrity at temperatures ranging from –80oC to 60oC. Suitable for packaging, ambient and cryogenic storage of a variety of liquids including media, buffers, process liquids, and biological products.