This item is now discontinued. We recommend the following as suggested replacements: WW-09411-60
Liberty Industries 6-301 Adhesive Roll Mop, Handheld, 9"W, with 20 Perforated Sheets
Representative image only.
Liberty Industries Mfr # 6-301 – Item # WW-09411-50
Quickly clean surfaces of dust and lint
  • Adhesive traps dirt and removes it easily
  • Simply peel back layer to expose clean sheet
  • Perforated rolls for easy tearing—each roll contains 20 sheets
SPECS & DESCRIPTION
Specifications & Description
  • Width (in)9
  • DescriptionAdhesive Roll Mop, handheld, 9\"W, with 20 perforated sheets
Key Features
  • Adhesive traps dirt and removes it easily
  • Simply peel back layer to expose clean sheet
  • Perforated rolls for easy tearing—each roll contains 20 sheets
More About this Item
Cut contamination with these adhesive roll mops. Mops have adhesive coated surfaces to remove debris and lint from glass, plastic, metal, and other problem surfaces. Choose from handheld or telescoping mops.
Handheld model 09411-40 is perfect for cleaning work counters, windows, or anywhere quick cleaning is needed in a confined space. Telescopic model 09411-60 has a 48"L handle that extends to 7 feet—ideal for hard to reach areas like walls, ceilings, floors, and corners.
Each roll mop has 20 disposable adhesive sheets; peel away the top layer to expose a fresh surface. Sheets are perforated every 14-3/8" for easy access to a fresh sheet. Refill rolls for each mop are sold separately.