Lascar Electronics EL-GFX-2+ Temperature and Humidity Data Logger with Graphic LCD Screen
Representative image only.
Lascar Electronics Mfr # EL-GFX-2+ – Item # WW-23039-14
Graphic LCD provides real-time information on data being recorded
  • Ability to start, stop, graph trends, and summarize data in the field without computer connection
  • Memory capacity exceeds 250,000 readings per channel with resolution to the nearest 0.1 digit
  • Windows®-based control software available as free download
  • Compact and robust waterproof design with IP67 rating
  • Wall-mounting clip for quick magnetic mounting to a metallic surface
  • 1 year warranty
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • Number of Channels1
  • Input TypeTemperature
  • Max Temperature (° F)257
  • Min Temperature (° F)-40
  • Max Temperature (° C)125
  • Min Temperature (° C)-40
  • Display TypeLCD
  • DescriptionTemperature and Humidity Data Logger with Graphic LCD Screen
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Ability to start, stop, graph trends, and summarize data in the field without computer connection
  • Memory capacity exceeds 250,000 readings per channel with resolution to the nearest 0.1 digit
  • Windows®-based control software available as free download
  • Compact and robust waterproof design with IP67 rating
  • Wall-mounting clip for quick magnetic mounting to a metallic surface