LaMotte BioPaddles 5551 Sabouraud Dextrose Agar Microbiological Test Kit
Representative image only.
LaMotte BioPaddles Mfr # 5551 – Item # WW-99532-44
Identify and quantify microbes in water and soil, or on any surface
  • Unique design enhances colony morphology characterization
  • Versatile kit can test via liquid, surface or air
  • No need for inoculating loops, Bunsen burners or refrigeration
  • Simple incubation requirements
PRODUCT OPTIONS
₹11,400.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestFungi, yeasts and molds
  • Reagent PlatformSabouraud Dextrose
  • DescriptionSabouraud Dextrose Agar Microbiological Test Kit
Key Features
  • Unique design enhances colony morphology characterization
  • Versatile kit can test via liquid, surface or air
  • No need for inoculating loops, Bunsen burners or refrigeration
  • Simple incubation requirements
More About this Item
BioPaddles are flexible, dual-agar paddles with microbe-specific media enclosed in a sterile vial. Identify and quantify microbes in air, soil, water or any surface! Biopaddles do not require any other testing equipment— only a magnifier and a warm place (35°C); incubator optional.
The LaMotte BioPaddle Colony Identification App is available to compare unkown colonies to known colony examples for the various media types. App also contains information regarding organisms, microhabitats, techniques, and more. Follow the instructions included with your Biopaddle to access the free app on any smart device.
99532-44 is specific for selective cultivation of fungi (yeasts and molds).