Laboratorio Electrofisco 60-B411756H Portable Gaussmeter w/ Axial Probe
Representative image only.
Laboratorio Electrofisco Mfr # 60-B411756H – Item # WW-20001-62
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPortable Gaussmeter w/ Axial Probe
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Package includes CC-4D Carrying Case, 6' Probe Cable, Batteries
and NIST Calibration Certificate.
FEATURES
High accuracy and resolution
Measure DC and AC fields; True RMS readings to 20 kHz with AC probes
Range extendable to 150 kG with select probes
Analog Output
High impact foam-lined plastic carrying case
Operates up to one year with standard “AA” alkaline batteries
APPLICATIONS
Measure residual fields
Analyze magnetic circuits and components
Classify magnets
Measure absolute and differential fields
Plot field uniformity
Measure stray and leakage fields