Labconco FlaskScrubber 4540031 Vantage Series Glassware Washer, Window And Interior Light, 230V

Representative image only.
Labconco FlaskScrubber Mfr # 4540031 – Item # WW-98964-50
Purified-water rinses and HEPA filtered forced-air drying eliminate soluble, airborne contaminants
  • Automatic detergent and rinse aid dispensing eliminates the need to fill manual dispensers
  • Ten factory preset cycles make operation easy
  • Accommodates and cleans a wide variety of glassware, including volumetric flasks
  • HEPA-filtered forced air drying removes 99.97% of 0.3µm particulates, reducing contaminants from adhering to your glassware
  • RS-232 port for downloading information for validation
  • Built-in alarms warn of hazardous conditions

SPECS & DESCRIPTION

Specifications & Description

  • Water Consumption (Gal)3.4
  • Exterior Width (in)32.25
  • Exterior Height (in)34.25
  • Exterior Depth (in)27.5
  • Chamber Width (in)22.625
  • Chamber Height (in)19
  • Chamber Depth (in)21.3125
  • Power (VAC)208 / 240
  • Power (Hz)50/60
  • Washing Temperature (° F)199
  • Washing Temperature (° C)93
  • Inlet ConnectionMunicipal hot water: 3/8\" NPT(F), Distilled water: 3/8\" hose barb
  • Outlet Connections3/4\" ID tubing
  • Shipping Weight (lbs)294
  • DescriptionVantage Series Glassware Washer, Window And Interior Light, 230V
Conformite Europeenne

Key Features

  • Automatic detergent and rinse aid dispensing eliminates the need to fill manual dispensers
  • Ten factory preset cycles make operation easy
  • Accommodates and cleans a wide variety of glassware, including volumetric flasks
  • HEPA-filtered forced air drying removes 99.97% of 0.3µm particulates, reducing contaminants from adhering to your glassware
  • RS-232 port for downloading information for validation
  • Built-in alarms warn of hazardous conditions

More About this Item

Thirty-six detachable spindles reach inside narrow-neck glassware for complete cleaning, and forced air dries the interior. LCD provides easy monitoring of wash cycle, including water temperature.
Ten factory preset cycles and two user programmable cycles allow you to customize your wash settings. Cycle start delay allows you to set unit to run up to 8 hours later to reduce unwanted noise and lower energy consumption during peak hours.
Two water pumps, one for washing and one for draining, reduce cross-contamination. Increased water temperature of 93°C (199°F) allows for glassware sanitization and better cleaning.
What's included washer with 304 stainless steel 36-spindle lower rack, eighteen glassware holders, thirty-six glassware clips, ten small spindles, automatic detergent dispenser, interior light, four leveling feet, and utility hookup terminals.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat