Labconco CApture Portable Fuming Station, 230 V

Representative image only.
Labconco Mfr # 3420020 – Item # WW-28616-10
Quickly fixes latent fingerprints at the crime scene
  • Unique collapsible design is easy to transport and set up
  • Uses microburst or traditional CA fuming methods
  • Protects the user from hazardous fumes without ducting
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Exterior Width (in)10.75
  • Exterior Height (in)19
  • Exterior Depth (in)15.5
  • Power (VAC)230
  • Power (Hz)50
  • DescriptionPortable Fuming Station, 230 V
  • Warranty1 Year
1 Year WarrantyConformite Europeenne

Key Features

  • Unique collapsible design is easy to transport and set up
  • Uses microburst or traditional CA fuming methods
  • Protects the user from hazardous fumes without ducting
  • 1 year warranty

More About this Item

Bring this portable fuming system directly to the crime scene, saving valuable laboratory time and space. System combines all the components needed for fingerprint fuming in one compact, easy-to-transport unit. A microprocessor-controlled heater heats the CA adhesive (Super Glue®). The heater may be set in one degree increments between 120 to 350°F (49 to 77°C). An LCD displays the current or set-point temperature. A shatter-resistant, clear polycarbonate protective chamber, 8" W x 16" H x 8" D (20 x 41 x 20 cm), contains the fume as they react with the latent prints. A built-in exhaust blower pulls air from the chamber, through the filter and exhausts it into the room. A particulate/carbon exhaust filter traps smoke and CA fumes exhausted from the chamber.


Constructed of durable epoxy-coated steel with a stainless steel manifold to support evidence inside the chamber. A stainless steel hook, shelf and perforated tray are included. The CApture system runs on 115 VAC, 60 Hz, , 230 VAC, 50 Hz, or can be run on car battery when connected to a user-supplied inverter.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat