Labconco 8000300 Carboy Caddy, 20 x 23-5/8" Platform
Representative image only.
Labconco 8000300 Carboy Caddy, 20 x 23-5/8" Platform
Labconco | Mfr: 8000300| Item: 0937805
Safe and effective way to move large carboys, drums, and crates!
₹141,000.00INR / Each
₹141,000.00INR / Each
Product Description
Key Features
  • Limited Lifetime Warranty
More About this Item
Helps eliminate back strain and heavy lifting; just tip cart forward and slide carboy on the cart. Square still tubing supports phenolic base with a melamine high pressure laminate surface. Hard rubber casters swivel 360° to easy navigation. Base is 4-1/4'' from the floor. Handle is 38-1/2''H x 19''W.
Limited Lifetime Warranty
Specifications
  • DescriptionCarboy Caddy, 20 x 23-5/8" Platform
  • WarrantyLimited Lifetime