Labconco 5220320 XPERT Weigh Box, 33.5" Wide; 230 VAC

Representative image only.
Labconco Mfr # 5220320 – Item # WW-34764-10
Ideal for safe and economical weighing and handling
  • Protection from hazardous airborn particulates
  • Anti-static ionizer fan to reduce static
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Transfer Chamber Width (in)25
  • Transfer Chamber Height (in)33.5
  • Transfer Chamber Width (cm)63.5
  • Transfer Chamber Height (cm)85.1
  • Transfer Chamber Length (in)27.5
  • Transfer Chamber Length (cm)69.9
  • Width of Equipment Sleeve (in)11
  • Width of Equipment Sleeve (cm)28
  • Chamber Width (in)33.5
  • Chamber Height (in)25
  • Chamber Depth (in)27.5
  • Exterior Width (in)52.7031
  • Exterior Height (in)31.6093
  • Exterior Depth (in)40
  • Chamber Width (cm)85.09
  • Chamber Height (cm)63.5
  • Chamber Depth (cm)69.85
  • Exterior Width (cm)133.9
  • Exterior Height (cm)80.3
  • Exterior Depth (cm)101.6
  • Power (VAC)230
  • Power (Hz)50
  • DescriptionXPERT Weigh Box, 33.5\" Wide; 230 VAC
  • Warranty1 year
1 Year WarrantyConformite EuropeenneISO9001 Certified Supplier

Key Features

  • Protection from hazardous airborn particulates
  • Anti-static ionizer fan to reduce static
  • 1 year warranty

More About this Item

These economical weigh boxes feature a liner of one-piece molded, medium density polyethylene and a dry powder epoxy-coated steel exterior. The chamber features a 1/4" thick laminated safety glass viewing window with powder-coated steel frame and neoprene gasket. The cutout is 26 2/5"W x 12 1/2"H, and is easily removable for loading equipment. Space-saving inner and outer transfer chamber (11" nominal ID x 12"L) doors pivot upward, and are counterbalanced and equipped with quick-latches. Units also feature an interior ceiling mounted electrical duplex receptacle with a 5 amp maximum load, and a flourescent lamp that is easily replaceable from the top of the unit. The black, chemical resistant work surface is sealed to the glove box floor, allowing for easy clean-up of spills
Units also feature both inlet and outlet HEPA filters and provide a physical barrier to protect the operator from hazardous airborne particles and powders, making these units ideal for use with pharmaceutical research, nanotechnology, biochemistry, Microorganisms, low-level radio-chemicals, chemical carcinogens and asbestos. Each unit features a built-in blower with airflow from 13 to 100 cubic feet per minute (CFM). The HEPA filters are 99.99% efficient on 0.3micron particulates. The main chamber static pressure gauge monitors HEPA filter loading. Airborne particulate cleanliness has been measured and exceeds ISO Class 5 conditions, and acheived ISO ISO Class 3 conditions at all airflow volumes. The anti-static ionizer fan circulates ionized air inside the box to reduce static charge. To minimize balance vibration, a 13 2/5"W x 20 2/5"D x 2.7"H marble slab with 304 stainless stell cover is included within the weigh box. These units have been factory tested with virtually no leaks detected.
Units require a supporting base, see accessories below for ordering information.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat