Labconco 4588500 Lower Standard Rack for Glassware Washers
Representative image only.
Labconco Mfr # 4588500 – Item # WW-98963-61
Optimize performance of your Labconco glassware washer
  • Adds more cleaning options to your washer
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionLower standard rack for Glassware Washers
  • Warranty1 year
1 Year Warranty
Key Features
  • Adds more cleaning options to your washer
  • 1 year warranty
More About this Item
Choose from a wide range of rack accessories and other products to ensure top performance of your Labconco glassware washer.
Lower standard rack is included with all SteamScrubber® glassware washers. Standard rack may be used in all Labconco glassware washers, including the FlaskScrubber® and FlaskScrubber® Vantage® models. Rack accommodates a variety of racks and inserts so that you can customize the washer to your glassware mix.