Labconco 4494200 Pipet holder, holds eight 1 to 10 mL pipets
Representative image only.
Labconco Mfr # 4494200 – Item # WW-98952-69
Optimize performance of your Labconco FlaskScrubber
  • Constructed of durable 304 stainless steel
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPipet holder, holds eight 1 to 10 mL pipets
  • Warranty1 year
1 Year Warranty
Key Features
  • Constructed of durable 304 stainless steel
  • 1 year warranty
More About this Item
Pipet holders permit direct injection cleaning of pipets. Self-sealing silicone check valves allow washing of partial or full loads. Installation hardware included.