Labconco 3746713 Telescoping Base Stand with Casters, 60"W X 29"D
Representative image only.
Labconco Mfr # 3746713 – Item # WW-09109-45
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Hood Width (in)60
  • Exterior Width (in)60
  • Exterior Height (in)29
  • DescriptionTelescoping Base Stand with Casters, 60\"W X 29\"D
  • Warranty1 Year
1 Year WarrantyISO9001 Certified Supplier
Key Features
  • 1 year warranty
More About this Item
Support Labconco workstations by using one or more base stands. Total width of stands must equal width of workstation. Adjust to seven height positions to accommodate sitting or standing.
Telescoping base stands feature -3/4" tubular steel frame with glacier white powder coating; include leveling feet. They are adjustable during installation in 1" increments from 27-1/2" to 34-1/2".