Labconco 3410021 Protector Downdraft Powder Station, 3-ft Wide, 230 V

Representative image only.
Labconco Mfr # 3410021 – Item # WW-28616-34
Fingerprint powdering of large items is no longer a challenge!
  • Safely work with nuisance powders
  • Open-side, open-top design allows unrestricted work space and accommodates oversized items
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Hood Width (in)36
  • Exterior Width (in)36
  • Exterior Height (in)13.3125
  • Exterior Depth (in)25.8125
  • Power (VAC)230
  • Power (Hz)50
  • Filter TypeHEPA
  • DescriptionProtector Downdraft Powder Station, 3-ft Wide, 230 V
  • Warranty1 Year
1 Year WarrantyConformite Europeenne

Key Features

  • Safely work with nuisance powders
  • Open-side, open-top design allows unrestricted work space and accommodates oversized items
  • 1 year warranty

More About this Item

The HEPA filtered, open design of the downdraft powder stations safely contain powders while giving the freedom of an open work area. With the addition of the optional transition connector 28616-38, multiple downdraft powder stations can be joined together to create a longer, continuous work station.

Durable workstations are constructed of epoxy coated steel and include a removable stainless steel work surface. Beneath the work surface is a powder collection tray that is removable for easy clean up. The 75-watt blower is adjustable to optimize the life of each HEPA filter. A filter alarm light alerts you to low airflow conditions prompting a filter change. Each downdraft powder station comes complete with a prefilter and a HEPA filter; available in 2-ft (0.6-m) and 3-ft (0.9-m) widths

Note This product is for nuisance powders only. It is not to be used for biohazards, toxic chemicals or radioisotopes.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat