Lab Armor 74220-714 Waterless bead bath 14L. with Beads 12L, 220V

Representative image only.
Lab Armor Mfr # 74220-714 – Item # WW-10122-15
Low maintenance, eco-friendly alternative to water baths
  • Eco-friendly — beads are recyclable, use 50% less electricity, and do not require germicides
  • Beads hold things in place, eliminating the need for racks and weights
  • Bath is always ready, eliminating warm-up times
  • Safer operation than oil baths
  • Dry, inert, nontoxic aluminum oxide ensures hazard-free operation emptying, cleaning, and refilling
  • 1 year warranty

₹603,000.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • Bath TypeBead Bath
  • Reservoir Capacity (L)14
  • Max Temperature (° C)80
  • Min Temperature (° C)Ambient +5
  • Temperature Uniformity (° C)±1.0
  • Width (in)15 7/8
  • Height (in)18 1/2
  • Depth (in)8 1/2
  • Bath Opening Width (in)12 1/8
  • Bath Opening Length (in)12
  • Immersion Depth (in)6
  • Temperature SettingDigital
  • Wetted MaterialsSS
  • Power (VAC)230
  • Power (Hz)50/60
  • Power (amps)2.5
  • DescriptionWaterless bead bath 14L. with Beads 12L, 220V
  • Warranty1 Year
1 Year WarrantyConformite Europeenne

Key Features

  • Eco-friendly — beads are recyclable, use 50% less electricity, and do not require germicides
  • Beads hold things in place, eliminating the need for racks and weights
  • Bath is always ready, eliminating warm-up times
  • Safer operation than oil baths
  • Dry, inert, nontoxic aluminum oxide ensures hazard-free operation emptying, cleaning, and refilling
  • 1 year warranty

More About this Item

These baths use metallic beads instead of water, delivering exceptional temperature uniformity of +/-1 degree without lengthy warm-up times. Beads create a dry bath, avoiding the contamination and maintenance of traditional water baths. They also improve energy efficiency over 50% of traditional water baths.

Beads hold a variety of containers securely in place without any float-away or ruined samples. You are no longer limited to water-tight containers, so you can incubate multi-well plates, petri dishes, and open-top samples at any angle.

Metallic beads have high thermal conductivity in the range of +/-180° C. Smooth, rounded surfaces measure 5 to 8 mm diameter and 1 to 2 mm high. Beads are moisture and gas impermeable, and are easy to keep clean without harmful germicides. These versatile beads can also be used in a container for freezers, ovens, and incubators.

Includes beads for bath.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat