Kruss DS7800 Benchtop Density Meter, 90 to 240 VAC, 50/60 Hz

Representative image only.
Kruss Mfr # DS7800 – Item # WW-25750-70
Integrated Peltier thermostat for excellent temperature control
  • Interface with RS-232, USB, and Ethernet for connection flexibility
  • Stores the last 999 measurements for data recall
  • 2 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Min Density Range (g/cm³)0
  • Max Density Range (g/cm³)1.9999
  • Specific Gravity Resolution0.0001 g/cm3
  • Max Temperature (° F)104
  • Min Temperature (° F)50
  • Max Temperature (° C)40
  • Min Temperature (° C)10
  • Sample Viscosity Max50 mPa x s
  • Sample Volume (mL)2
  • Power (VAC)90 to 240
  • DescriptionBenchtop Density Meter, 90 to 240 VAC, 50/60 Hz
Two year warranty

Key Features

  • Interface with RS-232, USB, and Ethernet for connection flexibility
  • Stores the last 999 measurements for data recall
  • 2 year warranty

More About this Item

Determine mixing ratios and perform quality and quantity inspection in the chemical, pharmaceuticals, cosmetics, food and beverage, pulp and paper, petrochemcial, and wastewater industries. Benchtop density meter measures using the oscillating tube technique: an oscillating U-tube is filled with the sample and the change in oscillation frequency and time is proportional to the density.

High-accuracy integrated Peltier thermostat provides excellent temperature stability. Other meter features include user-friendly touch screen with menu-driven operation, multiple computer interfaces (RS-232, USB, and Ethernet), and six language options (English, French, Italian, German, Portuguese, and Spanish).

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat