Koehler K22009 Viscometer Tube, Brass, for Saybolt Viscometer Bath
Representative image only.
Koehler Mfr # K22009 – Item # WW-98928-52
Optimize performance of your Koehler saybolt viscometer bath
  • Keep your saybolt viscometer bath running properly with these accessories
₹128,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialBrass
  • DescriptionViscometer Tube, Brass, for Saybolt Viscometer Bath
Key Features
  • Keep your saybolt viscometer bath running properly with these accessories
More About this Item
Choose from a wide range of replacement parts and accessories to ensure top performance of your Koehler saybolt viscometer bath.
Saybolt Viscometer Tube is precisely machined to meet ASTM requirements. Mount tubes vertically in viscometer bath with supplied mounting hardware. Conform to ASTM D88 and E102 standards.