Cole-Parmer Vial Plug Cap, 12mm, Polyethylene; 1000/pk
Representative image only.
Cole-Parmer Item # WW-98702-34
Optimize performance of your Kinesis shell vials
  • Polyethylene plugs give a tight seal yet are easy to insert or remove
PRODUCT OPTIONS
₹8,500.00INR / Pkg of 1000
SPECS & DESCRIPTION
Specifications & Description
  • Cap TypePlug
  • Cap Diameter (mm)12
  • Cap MaterialPolyethylene
  • SterileNo
  • Special FeaturesNA
  • DescriptionVial Plug Cap, 12mm, Polyethylene; 1000/pk
Key Features
  • Polyethylene plugs give a tight seal yet are easy to insert or remove
More About this Item
These shell vial plugs are compatible for use with the Kinesis Shell Vials. Choose from polyethylene or polyethylene plugs.