Cole-Parmer Vial and Cap Kit, MRQ30, Glass Vials with PTFE/Silicone Septa; 100/pk
Representative image only.
Cole-Parmer Item # WW-98706-70
Low residual volume—less than 1 µL!
  • Compatible with auto samplers
  • Clean enough for LCMS analysis
  • Comes complete with caps and septa for convenience
SPECS & DESCRIPTION
Specifications & Description
  • Tube MaterialGlass
  • Tube ColorClear
  • Cap TypeShort Thread
  • Cap Diameter (mm)9
  • Cap MaterialPolypropylene
  • Septum TypeSilicone/PTFE
  • SterileNo
  • Special FeaturesTotal Recovery
  • DescriptionVial and Cap Kit, MRQ30, Glass Vials with PTFE/Silicone Septa; 100/pk
Key Features
  • Compatible with auto samplers
  • Clean enough for LCMS analysis
  • Comes complete with caps and septa for convenience
More About this Item
These MRQ30 total recovery vials deliver maximum sample capacity with minimal residual volume (<1 µL). Vials have 9-mm short thread that sits much higher on the vial allowing you to screw on the caps and still use them in a robotic autosampler. Wide-mouth opening provides a large target area reducing the possibility of bent needles—use with bottom or side draw-port needles.
Vials are made of Type 1, Class A 33-expansion borosilicate glass to give you the highest resistance to chemical attack. Low bleed makes these vials clean enough for LCMS analysis.