Cole-Parmer TELOS® SLE Supported Liquid Extraction Wells, Neutral Matrix 545E, 50 mg; 100/PK
Representative image only.
Cole-Parmer Item # WW-06482-03
Perfectly suited for automation and high throughput procedures
  • Efficient extraction
  • High analyte recovery with no emulsion formation
  • Loose wells
  • Neutral matrix (NM) 545E
  • Requires base plate 06474-60
PRODUCT OPTIONS
₹32,600.00INR / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • Bed Mass (mg)50
  • ApplicationsNeutral matrix (NM) 545E
  • DescriptionSLE Supported Liquid Extraction Wells, Neutral Matrix 545E, 50 mg; 100/PK
Key Features
  • Efficient extraction
  • High analyte recovery with no emulsion formation
  • Loose wells
  • Neutral matrix (NM) 545E
  • Requires base plate 06474-60
More About this Item
Use supported liquid extraction plates when you need to extract samples quickly and effectively. Wells are packed with a diatomaceous earth, a neutral matrix, to provide support to absorb the aqueous sample. Once the aqueous sample is fully absorbed, a water immiscible organic solvent is used to elute the analytes. High analyte recoveries are obtained without any offline steps such as protein precipitation, and sample extracts are free of proteins and phospholipids. A hydrophilic top frit allows the sample to pass through while minimizing any blockage.
NM 545E uses a larger particle size material for easier flow. Use the loose wells when the sample numbers vary, populating the base plate with only the desired number of wells. Base plate 06474-60 is required to use loose wells.