Cole-Parmer TELOS® PPT Protein Precipitation 96-Well Plate; 1/EA
Representative image only.
Cole-Parmer Item # WW-06482-28
Efficient and reliable protein precipitation by filtration
  • Ideal for high-thoughput applications
₹12,100.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells96
  • Plate ColorTranslucent
  • Binding PropertyProtein
  • DescriptionTELOS® PPT Protein Precipitation 96-Well Plate; 1/EA
Key Features
  • Ideal for high-thoughput applications
More About this Item
Clean up biological fluid samples, especially plasma, using TELOS PPT protein precipitation plate. Complete protein precipitation process takes place in the well—no offline liquid handling and centrifugation steps are required. Specially designed double frits provide fast and trouble-free protein precipitation. One frit holds up the precipitation solvent, acetonitrile, while the other frit with larger porosity prevents blocking, allowing reproducible collection of the subsequent filtrate.
Dispense sample directly into the acetonitrile with force to achieve reproducible precipitation from sample to sample. The optimum ratio of acetonitrile to plasma is 4:1 v/v. This ensures complete precipitation occurs and the filtrate is precipitate free.