Cole-Parmer TELOS® Polar SPE Column, MgO3Si, 500 mg sorbent, 3 mL; 50/pk
Representative image only.
Cole-Parmer Item # WW-06476-33
Extract polar analytes from nonpolar organic solvents
  • Commonly used for separation of lipids and pesticide residue extraction
  • Compatible with manual and automated sample processing protocols
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • Bed Mass (mg)500
  • Column Particle Size (µm)125
  • Wetted MaterialsPolypropylene body and polyethylene frits
  • Total Volume Capacity (mL)3
  • SorbentFlorisil® PR Grade
Key Features
  • Commonly used for separation of lipids and pesticide residue extraction
  • Compatible with manual and automated sample processing protocols
More About this Item
Use the TELOS polar (normal phase) SPE columns to retain the analytes of interest or to remove polar interferences from the sample. Columns with Florisil® PR grade (magnesium silicate) sorbent have larger particle size than silica and NH2. Florisil has a pore diameter of 70Å. All columns are made of polypropylene (except 06476-29, made of glass) and feature Luer-slip outlet.