Kinesis TELOS® Guard Cartridge, Silica HILIC, 5 µm, 4.6 mm ID; 5/PK
Representative image only.
Kinesis Item # WW-06481-42
Help extend the life of your columns
  • Optimize performance of your Kinesis® TELOS® HPLC and UHPLC columns
  • Packed with silica HILIC
SPECS & DESCRIPTION
Specifications & Description
  • Compatible With4.6 mm ID silica HILIC columns
  • DescriptionTELOS® Guard Cartridge, Silica HILIC, 5 µm, 4.6 mm ID; 5/PK
Key Features
  • Optimize performance of your Kinesis® TELOS® HPLC and UHPLC columns
  • Packed with silica HILIC
More About this Item
Support your columns with optional guard cartridges (and holder), available packed with identical packing material.