Kinesis TELOS® Guard Cartridge, C18, 5 µm, 4.6 mm ID; 5/PK
Representative image only.
Kinesis Item # WW-06481-95
Help extend the life of your columns
  • Optimize performance of your Kinesis® TELOS® HPLC and UHPLC columns
  • Packed with C18
₹73,500.00INR / Pkg of 5
SPECS & DESCRIPTION
Specifications & Description
  • Compatible With4.6 mm ID C18 columns
  • DescriptionTELOS® Guard Cartridge, C18, 5 µm, 4.6 mm ID; 5/PK
Key Features
  • Optimize performance of your Kinesis® TELOS® HPLC and UHPLC columns
  • Packed with C18
More About this Item
Support your columns with optional guard cartridges (and holder), available packed with identical packing material.