Kinesis TELOS Flash Chromatography Column, Luer-lok and slip tip, Silica, 25 g; 10/pk
Representative image only.
Kinesis Item # WW-06474-73
Use with systems from ISCO® and Buchi®
  • Optimized column geometry provides maximum recovery
  • Excellent for purification of a wide range of synthetic and natural compounds
  • Use with Reveleris® from Buchi® and Flash systems from ISCO® and Interchim®
SPECS & DESCRIPTION
Specifications & Description
  • Bed Mass (g)25
  • Column Particle Size (µm)50
  • Wetted MaterialsPolypropylene column, polyethylene frit
  • Max Pressure (PSI)180
  • SorbentSilica*
Key Features
  • Optimized column geometry provides maximum recovery
  • Excellent for purification of a wide range of synthetic and natural compounds
  • Use with Reveleris® from Buchi® and Flash systems from ISCO® and Interchim®
More About this Item
Make connections to flash chromatography instruments quick and easy with Luer connectors. Columns are made of polypropylene and have pressure ratings to support common flash chromatography back pressures.