Cole-Parmer TELOS® Flash Chromatography Column, Luer-lok and slip tip, NH2, 120 g; 5/pk
Representative image only.
Cole-Parmer Item # WW-06475-04
Use with systems from ISCO® and Buchi®
  • Optimized column geometry provides maximum recovery
  • Use to separate primary, secondary, and tertiary amines
  • NH2 shows better peak shapes with polar and basic compound—ideal for carbohydrates
PRODUCT OPTIONS
₹134,000.00INR / Pkg of 5
SPECS & DESCRIPTION
Specifications & Description
  • Bed Mass (g)120
  • Column Particle Size (µm)50
  • Wetted MaterialsPolypropylene column, polyethylene frit
  • Max Pressure (PSI)180
  • SorbentNH2
Key Features
  • Optimized column geometry provides maximum recovery
  • Use to separate primary, secondary, and tertiary amines
  • NH2 shows better peak shapes with polar and basic compound—ideal for carbohydrates
More About this Item
Make connections to flash chromatography instruments quick and easy with Luer connectors. Columns are made of polypropylene and have pressure ratings to support common flash chromatography back pressures.