Cole-Parmer TELOS® Flash Chromatography Column, Luer lock, NH2, 120 g; 5/pk
Representative image only.
Cole-Parmer Item # WW-06475-03
Use with SP and Idolera™ systems from Biotage®
  • Optimized column geometry provides maximum recovery
  • Use to separate primary, secondary, and tertiary amines
  • NH2 shows better peak shapes with polar and basic compound—ideal for carbohydrates
SPECS & DESCRIPTION
Specifications & Description
  • Bed Mass (g)120
  • Column Particle Size (µm)50
  • Wetted MaterialsPolypropylene column, polyethylene frit
  • Max Pressure (PSI)180
  • SorbentNH2
Key Features
  • Optimized column geometry provides maximum recovery
  • Use to separate primary, secondary, and tertiary amines
  • NH2 shows better peak shapes with polar and basic compound—ideal for carbohydrates
More About this Item
Make connections to flash chromatography instruments quick, easy, and secure with Luer-lok connectors. Columns are made of polypropylene and have pressure ratings to support common flash chromatography back pressures.