Cole-Parmer TELOS® Endcapped Nonpolar SPE Microplate, C18 AQ, 5 mg sorbent, populated plate; 1/pk
Representative image only.
Cole-Parmer Item # WW-06476-94
Comes with 96 wells already in place yet allow the flexibility to remove any unwanted wells
  • Optimized for sample processing and extraction of small volume biological fluids
  • Use with vacuum or positive pressure
  • Applications: Nonpolar
SPECS & DESCRIPTION
Specifications & Description
  • Bed Mass (mg)5
  • Carbon Loading18%
  • Column Particle Size (µm)50
  • End CapYes
  • ApplicationsNonpolar
  • SorbentC18 (AQ)
Key Features
  • Optimized for sample processing and extraction of small volume biological fluids
  • Use with vacuum or positive pressure
  • Applications: Nonpolar
More About this Item
The internal diameter, frit design, and sorbent material of the Cole-Parmer® TELOS® Silica-Based SPE populated-well microplates allow for maximum recovery with minimal sample dilution and reduced evaporation time. Select endcapped sorbents for applications requiring minimal secondary silanol interactions to enhance compound retention and elution. Use C18 (AQ) with samples with large volumes of water.