Cole-Parmer TELOS® Endcapped Nonpolar SPE Column, C2, 100 mg sorbent, 1 mL; 100/pk
Representative image only.
Kinesis Item # WW-06477-28
Extract hydrophobic or polar organic analytes from aqueous matrices
  • Features C2 sorbent
  • Applications: Extraction of hydrophobic and polar analytes
SPECS & DESCRIPTION
Specifications & Description
  • Bed Mass (mg)100
  • Carbon Loading6%
  • Column Particle Size (µm)50
  • End CapYes
  • Total Volume Capacity (mL)1
  • ApplicationsExtraction of hydrophobic and polar analytes
  • SorbentC2
Key Features
  • Features C2 sorbent
  • Applications: Extraction of hydrophobic and polar analytes
More About this Item
TELOS nonpolar (reversed phase) SPE columns are available in a wide range of sorbents for extraction of hydrophobic and polar analytes. All columns are endcapped making them ideal for applications requiring minimal secondary silanol interactions to enhance compound retention and elution. Columns are made of polypropylene and feature Luer-slip outlet. TELOS C2 has a pore size of 70 Å and average particle size of 50 µm.