Cole-Parmer Snap Cap, Polyethylene, White, 11 mm, Rubber/PTFE; 1000/pk
Representative image only.
Kinesis Item # WW-98706-52
No need for tool with quick and easy snap caps
  • Snap caps are ideal for use when a crimping tool is not available
  • Constructed of durable polypropylene or polyethylene
  • Select from a variety of caps and septa materials that fit your application and needs
PRODUCT OPTIONS
₹33,400.00INR / Pkg of 1000
SPECS & DESCRIPTION
Specifications & Description
  • Cap TypeSnap
  • Cap Diameter (mm)11
  • Cap MaterialPolyethylene
  • Cap colorWhite
  • Septum TypeRubber/PTFE
  • SterileNo
  • DescriptionSnap Cap, Polyethylene, White, 11 mm, Rubber/PTFE; 1000/pk
Key Features
  • Snap caps are ideal for use when a crimping tool is not available
  • Constructed of durable polypropylene or polyethylene
  • Select from a variety of caps and septa materials that fit your application and needs
More About this Item
Snap Top Caps can be snapped on and off by hand which eliminates the need for metal crimping and decapping tools. Snap caps are available in polypropylene (PP) or polyethylene (PE) with various septum materials. Select septa with slit to prevent a vacuum from forming inside the vials.
Caps are available in different colors, septum materials, and open or closed configurations to suit your application.