Cole-Parmer KX+ Vial and Cap Kit, 0.3 mL, Screw Top Glass Vials, PTFE/Silicone Septa, Mass Spec Quality; 100/pk
Representative image only.
Cole-Parmer Item # WW-98701-99
Super clean for highly sensitive GCMS and LCMS applications
  • Vials and caps are lot tested by LCMS for cleanliness—certificate of analysis included, complete with MS scan
  • Minimal pH shift—vials and closures have a neutral pH factor
  • Compatible with all autosamplers
  • Clear glass vials with PTFE/silicone septa
  • Made of Type I, Class A 33-expansion borosilicate glass
PRODUCT OPTIONS
₹24,900.00INR / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)0.3
  • Tube MaterialType I, Class A 33-expansion borosilicate glass
  • Tube Bottom ShapeConical
  • Tube ColorClear
  • Cap TypeScrew
  • Cap Diameter (mm)9
  • Cap MaterialPolypropylene
  • ThreadShort thread
  • Septum TypePTFE/silicone
  • SterileYes
  • Height (mm)32
  • Outside Diameter (mm)12
  • DescriptionKX+ Vial and Cap Kit, 0.3 mL, Screw Top Glass Vials, PTFE/Silicone Septa, Mass Spec Quality; 100/pk
Key Features
  • Vials and caps are lot tested by LCMS for cleanliness—certificate of analysis included, complete with MS scan
  • Minimal pH shift—vials and closures have a neutral pH factor
  • Compatible with all autosamplers
  • Clear glass vials with PTFE/silicone septa
  • Made of Type I, Class A 33-expansion borosilicate glass
More About this Item
LCMS certified vials are manufactured using a process that reduces surface activity, improving analytical quantification and minimizing pH shifts in the samples. Surface metals such as sodium and boron that can contaminate samples and interfere with trace analysis are effectively removed. Screw vials feature 9-mm short thread with a unique thread design that allows you to screw on the caps and still use them in a robotic autosampler. The thread sits much higher on the vial allowing space between the bottom of the thread and the shoulder of the vial.
Fused-insert vials are ideal for handling small-volume samples while delivering low residual volume (<8 µL).