Kinesis KX Membrane Filter, Hydrophilic PVDF, 25 mm dia., 0.45 µm; 100/pk
Representative image only.
Kinesis Item # WW-12918-07
Remove particulate without adversely affecting your final product
  • Use membrane filtration for samples holding suspended solids
  • Highly efficient filtration of particles according to size
  • Complete sample filtration with the least sample loss and energy use
  • Filtration for HPLC sample preparation and mobile phase
  • Uniform manufacturing for repeatable results
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • MaterialHydrophilic PVDF
  • Pore Size (µm)0.45
  • Diameter (mm)25
  • DescriptionMembrane Filter, Hydrophilic PVDF, 25 mm dia., 0.45 µm; 100/pk
Key Features
  • Use membrane filtration for samples holding suspended solids
  • Highly efficient filtration of particles according to size
  • Complete sample filtration with the least sample loss and energy use
  • Filtration for HPLC sample preparation and mobile phase
  • Uniform manufacturing for repeatable results