Kinesis EPA Septa, 24mm, Silicone natural/PTFE Beige; 1000/pk
Representative image only.
Cole-Parmer Item # WW-98700-75
Meets EPA requirements for testing environmental contaminants
  • Septa meet the requirements of the US Environmental Protection Agency (EPA)
  • Caps are constructed of durable white polypropylene
  • Select caps with UltraBond® septum to avoid sample contamination
  • Designed with a silicone/PTFE septa for the cleanest solution for contaminant free sample handling
  • Cap sold separately
PRODUCT OPTIONS
₹45,800.00INR / Pkg of 1000
SPECS & DESCRIPTION
Specifications & Description
  • Cap Diameter (mm)24
  • Septum TypeSilicone Natural/PTFE Beige
  • SterileNo
  • Special FeaturesSepta only
  • DescriptionEPA Septa, 24mm, Silicone Natural/PTFE Beige; 1000/pk
Key Features
  • Septa meet the requirements of the US Environmental Protection Agency (EPA)
  • Caps are constructed of durable white polypropylene
  • Select caps with UltraBond® septum to avoid sample contamination
  • Designed with a silicone/PTFE septa for the cleanest solution for contaminant free sample handling
  • Cap sold separately
More About this Item
These white polypropylene (PP) caps and EPA septa are free of substances that might influence sample analysis for testing of potentially harmful environmental contaminants in water or soil samples. Select from septa only, cap only, or cap and septa combination.