Cole-Parmer Deuterium (D2) Detector Lamp for Waters Acquity HB, Long Life
Representative image only.
Cole-Parmer Item # WW-11958-00
Economical replacement deuterium lamps save you money
  • Select from standard or long life lamps
  • Manufactured in an ISO-accredited Facility
  • Individually Tested and Issued with a Test Certificate
  • Long life 2000 hour bulb
  • Equivalent to Waters 201000281
  • 1 year warranty
PRODUCT OPTIONS
₹121,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithWaters Acquity HB
  • DescriptionDeuterium (D2) Detector Lamp for Waters Acquity HB, Long Life
  • Warranty1 year
1 Year Warranty
Key Features
  • Select from standard or long life lamps
  • Manufactured in an ISO-accredited Facility
  • Individually Tested and Issued with a Test Certificate
  • Long life 2000 hour bulb
  • Equivalent to Waters 201000281
  • 1 year warranty
More About this Item
Our range of deuterium (D2) lamps are made to fit the majority of detectors used in analytical labs today. Our lamps are used by many of the major pharmaceutical and contract research organizations (CROs) around the globe.
Our lamps are manufactured under stringent ISO 9001 quality procedures. Each lamp is manufactured to rigorous standards and traceable from manufacturing to customer delivery.
Works with Waters Acquity 2489 dual wavelength UV, 2998 PDA, Acquity, Acquity UPLC PDA, Acquity UPLC TUV, Acquity eLambda PDA.