Cole-Parmer Deuterium (D2) Detector Lamp for Hitachi U1800, U2800, U2810
Representative image only.
Cole-Parmer Item # WW-11941-72
Economical replacement deuterium lamps save you money
  • Select from standard or long life lamps
  • Manufactured in an ISO-accredited Facility
  • Individually Tested and Issued with a Test Certificate
  • Standard life 1000 hour bulb
  • 6 month warranty
PRODUCT OPTIONS
₹123,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithHitachi U1800, U2800, U2810
  • DescriptionDeuterium (D2) Detector Lamp for Hitachi U1800, U2800, U2810
  • Warranty6 month
6 Month Warranty
Key Features
  • Select from standard or long life lamps
  • Manufactured in an ISO-accredited Facility
  • Individually Tested and Issued with a Test Certificate
  • Standard life 1000 hour bulb
  • 6 month warranty
More About this Item
Our range of deuterium (D2) lamps are made to fit the majority of detectors used in analytical labs today. Our lamps are used by many of the major pharmaceutical and contract research organizations (CROs) around the globe.
Our lamps are manufactured under stringent ISO 9001 quality procedures. Each lamp is manufactured to rigorous standards and traceable from manufacturing to customer delivery.