Kinesis 96-well Collection Plate, Glass Lined PP, Flat Bottom, 2.4 mL; 10/pk
Representative image only.
Kinesis Item # WW-98700-11
Glass lining provides excellent chemical resistance
  • Constructed of glass-lined polypropylene
  • 96-well, flat bottom, 2.4 mL
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells96
  • Well Volume (µL)2400
  • Well ShapeRound, flat bottom
  • MaterialGlass-lined polypropylene
  • Description96-well Collection Plate, Glass Lined PP, Flat Bottom, 2.4 mL; 10/pk
Key Features
  • Constructed of glass-lined polypropylene
  • 96-well, flat bottom, 2.4 mL
More About this Item
Collection plates are available with three well shapes: V-bottom wells let you completely retrieve well content; U-bottom wells optimize coating and washing; and flat wells maximize light transmission.