Kimtech HC61170-56890 Sterile Critical Nitrile Gloves, small, bag of 20 pair
Representative image only.
Kimtech Mfr # HC61170-56890 – Item # WW-86486-00
Hand specific gloves are individually pair packed
  • Enhanced natural grip
  • Powder-free
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNitrile
  • Thickness (mil)5
  • SizeSmall
  • Length (in)12
  • Length (cm)30.5
  • ColorWhite
  • GripTextured
  • ComplianceISO 11137
  • SterileYes
  • DescriptionSterile Critical Nitrile Gloves, small, bag of 20 pair
Key Features
  • Enhanced natural grip
  • Powder-free
More About this Item
Lightly textured, powder-free white nitrile gloves feature a beaded cuff for easier donning. Lightly textured palms and fingertips enhance natural grip. Gloves are hand specific with a winged thumb for better fit and mobility. Gloves are sterilized via gamma irradiation and individually pair packed in a double polybag.