This item is now discontinued. We recommend the following as suggested replacements: WW-34523-38
DWK Life Sciences (Kimble) 15040G-30 Glass Bottle with Dropper, 30 mL, 12/cs
Representative image only.
DWK Life Sciences (Kimble) Mfr # 15040G-30 – Item # WW-16602-06
Heat and chemically resistant borosilicate glass
  • Autoclavable; except rubber nipples
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Volume (mL)30
  • Wetted PartsBorosilicate glass
  • DescriptionGlass Bottle with Dropper, 30 mL, 12/cs
  • Warranty1 year
1 Year Warranty
Key Features
  • Autoclavable; except rubber nipples
  • 1 year warranty
More About this Item
Dropper bottles are used in analytical, QC and environmental labs. Amber dropper bottles protect light sensitive chemicals, and are made to ASTM E438 specifications. Replacement droppers both glass and plastic are available below.